Order Entry
Spain
Orders LinkContactUsLinkComponent
Supelco® Organic Reference Standard, BTEX Solution

Supelco® Organic Reference Standard, BTEX Solution

BTEX Solution, Analytical standard, 1 mL
Catalog # 43728-1ML
Supplier:   Merck
Supelco® Organic Reference Standard, BTEX Solution
Catalog # 43728-1ML
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, BTEX Solution

BTEX Solution, Analytical standard, 1 mL

CAS Number:  
Price
71,80 €
Log in for availability
1 * 1 mL
Product Details & Documents
  • GHS signal: GHS08

Caution: P201 - P202 - P280 - P308 + P313 - P405 - P501

Documents

Specifications

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • Description:
    BTEX Solution
  • Cat. No.:
    43728-1ML
  • M-Clarity quality level:
    MQ100
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H340 - H350
  • Flash point:
    Not applicable
  • Density:
    1,099 g/ml at 20 °C
  • Shipping conditions:
    STD

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.