Ethyl oleate
Glass bottle for solids, 50 mg
CAYM10008201-50
: Elaidic acid ethyl ester, (Z)-9-Octadecenoic acid ethyl ester, Oleic acid ethyl ester
- :111-62-6
- :C₂₀H₃₈O₂
- :MFCD00009579
- :310.52 g/mol
- :12,06965
- :386 °C (1013 hPa)
- :–32 °C
- :0.87 g/cm³ (20 °C)
- :174 °C
- :Ambient
- :203-889-5
- Pack type:Glass bottle for solids
- Pk:50 mg