BioSpectrum 615, MOTORISED zoom f/2.8, MANUAL lift: 615 camera: 3.2 mp, 16 bit monochrome, 16 File Bit Depth (A/D), digital camera a nd mount, with binning capability (3.2 Mp). Peltier cooling to -60C from ambient. Camera mount and f/2.8 MOTORISED z 1 * 1 items
Catalog # UVPA97-0440-02
Supplier: UVP ULTRA VIOLET PRODUCTS
Frequently Bought Together