BioSpectrum 615, MOTORISED fixed 30mm, f/1.4, MOTORISED Lift: 615 camera: 3.2 mp, 16 bit monochrome, 16 File Bit Depth (A/D), digita l camera and mount, with binning capability (3.2 Mp). Peltier cooling to -60C from ambient. Camera mount and 30mm, f 1 * 1 items
Catalog # 733-2022
Supplier: UVP ULTRA VIOLET PRODUCTS
Frequently Bought Together