BioSpectrum 410 System. With Chemi HR camera: 12-bit monochrome, 16 File Bit Depth (A/D), digital camera and mount, 1600 x 1200 chip with binning capabilities, two stage peltier cooling to -50C from ambient, 24-70mm, f/2.8 MOTORISED zoom lens, UV fi 1 * 1 items
Catalog # UVPA97-0365-02
Supplier: UVP ULTRA VIOLET PRODUCTS
Frequently Bought Together