Order Entry
Spain
Orders LinkContactUsLinkComponent
Accessories for heating circulators
Accessories for heating circulators
Displacement insert W×D×H: 85×100×133 mm, CC-205B
Catalog # HUBE6041
Supplier:   HUBER
Accessories for heating circulators
Catalog # HUBE6041
Supplier:  HUBER
Accessories for heating circulators
Accessories for heating circulators
Displacement insert W×D×H: 85×100×133 mm, CC-205B
Price
420,00 €
Log in for availability
1 * 1 UN
Product Details & Documents
Specifications
  • Description:
    Displacement insert W×D×H: 85×100×133 mm
  • Seq:
    01
  • For:
    CC-205B