Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer), VWR®
TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer), Glass bottle, 2,5 L
Supplier: PROLABO CMP
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer), VWR®
Catalog # 87476.320
Supplier: PROLABO CMP
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Description:TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer)



