TUBING SET WITH SILICON LID F.SMART DIL. 1 * 1 items
Catalog # IULA2825
Supplier: IUL Instruments SA
TUBING SET WITH SILICON LID F.SMART DIL. 1 * 1 items
Catalog # IULA2825
Supplier: IUL Instruments SA
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Accessories for Mettler balances
Supplier: Mettler - Toledo
Titration beakers
Supplier: Mettler - Toledo
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Hand Tally Counter
Supplier: VWR Collection
Accessories for cell counter, EVE™
Supplier: NANOENTEK
VWR® Dehydrated Culture Media, Powdered
Supplier: VWR Chemicals
VWR® PH 20 pH Meter, Handheld
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR® pHenomenal® Electrode Stand
Supplier: VWR Collection
pH electrodes, SenTix® PLUS
Supplier: WTW
ThermoMixer® C Thermomixer
Supplier: EPPENDORF
VWR® Facial Tissues
Supplier: VWR Collection
TUBING FOR METALLIC EDGE SMART DILUTOR 1 * 1 items
Supplier: IUL Instruments SA
Flow Cells for MinION™ and GridION™ Devices
Supplier: Oxford Nanopore Technologies
Plastic cuvette for LICO
Supplier: Hach