Order Entry
Spain
Orders LinkContactUsLinkComponent
PFA inert sample introduction kit supplies
PFA inert sample introduction kit supplies
End cap for inert kit, PFA, with makeup gas port, suits inert sample introduction kit with Agilent 7500/7700/8800 series ICP-MS
Catalog # HEWLG3285-80020
Supplier:   Agilent
PFA inert sample introduction kit supplies
Catalog # HEWLG3285-80020
Supplier:  Agilent
PFA inert sample introduction kit supplies
PFA inert sample introduction kit supplies
End cap for inert kit, PFA, with makeup gas port, suits inert sample introduction kit with Agilent 7500/7700/8800 series ICP-MS
Price
754,00 €
Log in for availability
1 * 1 UN
Product Details & Documents
Inert kit for ICP-MS enables analysis of high-purity and HF-containing samples using a PFA-based sample intro system with inert Pt or sapphire-injector torch.
  • The O-ring free design and PFA construction material ensure low Background Equivalent Concentrations (BEC) and Detection Limits (DL)
  • Added flexibility for organic solvent analysis with an optional connecting pipe with gas port for the addition of O₂/Ar
  • Compatible with PFA MicroFlow concentric nebulizer, PFA MicroFlow nebuliser with I-AS probe, PFA concentric 200 μl/min nebulizer and PFA concentric 200 µl/min nebulizer with probe
They manufacture a range of high quality ICP-MS replacements, accessories and consumables including PFA Inert Sample Introduction Kit Supplies. A full range of accessories are available including connector and drain tubes, injector assemblies and torches for 7500 and 7700 ICP-MS systems. To maintain system performance, ensure that consumables are periodically replaced in accordance with recommended maintenance schedules.
A complete kit containing a PFA end cap, PFA spray chamber, connecting tube, demountable quartz torch, torch injector (four options are available: platinum or sapphire with a 2,5 mm or 1,5 mm injector) with PFA base. Easy to install in the standard Peltier-cooled spray chamber mount of the 7700. System uses a simple push-fit torch connector to aid assembly and disconnection
Specifications
  • Description:
    End cap for inert kit, PFA, with makeup gas port, suits inert sample introduction kit with Agilent 7500/7700/8800 series ICP-MS
  • Seq:
    02