Order Entry
Spain
Orders LinkContactUsLinkComponent
Silver nitrate ≥99%

Silver nitrate ≥99%

500 g
Catalog # A13854.36
Oxidizing
Corrosive
Environment
Silver nitrate ≥99%
Catalog # A13854.36
CAS Number:  7761-88-8
Silver nitrate ≥99%
Silver nitrate ≥99%

500 g

CAS Number:  7761-88-8
Price
1 140,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Lunar caustic, Silver mononitrate, Nitric acid silver salt
  • CAS Number:7761-88-8
  • Formula:AgNO₃
  • MDL Number:MFCD00003414
  • Molecular Weight:169.87 g/mol
  • Merck Index:12,08661
  • Boiling Pt:444 °C (1013 hPa)
  • Melting Pt:210 °C
  • Density:4.35 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-853-9
  • UN:1493
  • ADR:5.1,II
  • REACH:01-2119513705-43
Specifications

Specifications

  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.