ChemiDoc-It 615 Cam/Dkrm Kit: 615 camera: 3.2 mp, 16 bit monochrome, 16 File Bit Depth (A/D), digital camera and mount, with binning capability. Peltier cooling to -60C from ambient, MANUAL fixed 25mm, f/0.95 lens, close up diopter, USB 2.0 connect 1 * 1 items
Catalog # UVPA97-0270-02
Supplier: UVP ULTRA VIOLET PRODUCTS
Frequently Bought Together