EDGE FOR SEPTUM BAGS SMALL 1 * 1 ST
Catalog # IULA7528
Supplier: IUL Instruments SA
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® Culture Media, Ready to Use, in Bags
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Labelling tapes, temperature resistant
Supplier: PRECISION DYNAMICS TIMEM
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR® Culture Media, Ready To Use, In Plates
Supplier: VWR Chemicals
EDGE OF DISPENSING TUBE INOX 1 * 1 items
Supplier: IUL Instruments SA
VWR®, Accessories for FlexiPump® Peristaltic Pump
Supplier: VWR Collection