Order Entry
Spain
ContactUsLinkComponent
Ac-asp-glu-val-asp-7-amino-4-trifluoromethylcoumarin. A fluorogenic substrate for caspase-3 (cpp32, yama). Also acts as a substrate for granzyme b-processed caspase-6, caspase-7, caspase-8, and caspase-10. 1 * 10 mg
Catalog # ICNA03AFC13810
Supplier:  MP Biomedicals
Ac-asp-glu-val-asp-7-amino-4-trifluoromethylcoumarin. A fluorogenic substrate for caspase-3 (cpp32, yama). Also acts as a substrate for granzyme b-processed caspase-6, caspase-7, caspase-8, and caspase-10. 1 * 10 mg
Catalog # ICNA03AFC13810
Supplier:  MP Biomedicals
Frequently Bought Together