Diethyl ether, extra dry 99.5% stabilised, AcroSeal®
Glass bottle, 100 mL
Supplier: Thermo Fisher Scientific
Diethyl ether, extra dry 99.5% stabilised, AcroSeal®
Catalog # 326861000.
Supplier: Thermo Fisher Scientific
CAS Number: 60-29-7
Jump to:
Product Details & Documents
Synonyms: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
- CAS Number:60-29-7
- Formula:(CH₃CH₂)₂O
- MDL Number:MFCD00011646
- Molecular Weight:74.12 g/mol
- Merck Index:12,03852
- Boiling Pt:34.6 °C (760 mmHg)
- Melting Pt:–116 °C
- Density:0.714 g/cm³
- Flash Pt:–45 °C
- Storage Temperature:Ambient
- EINECS:200-467-2
- UN:1155
- ADR:3,I
- REACH:01-2119535785-29
Documents
Specifications
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.005 % (Coulometric)
- GC:≥99.4 %
- Stabilizer:1.5 to 10 ppm BHT
- Residue after evaporation:≤0.001 %
- Acidity:≤0.0002 meq/g
- Filt.over 0.2 micron PFTE:Filtered
- Peroxides:≤0.002 % (as H2O2)
Specifications
- Pack type:Glass bottle
- Pk:100 mL
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



