Probes for 845
Probe, paddle for measuring hardly accessible zones, TC type K, 0...300 °C, Class 2 (EN 60584-2), 145 mm, 8×145 mm
Catalog # 620-2253
Supplier: Testo
Jump to:
Product Details & Documents
Specifications
- Range:0...300 °C
- Description:Probe, paddle for measuring hardly accessible zones, TC type K
- Length:145 mm
- Seq:09
- Accuracy:Class 2 (EN 60584-2)
- Application:Surface
- Ø×L:8×145 mm