Order Entry
Spain
Orders LinkContactUsLinkComponent
Graduated pipettes, type 2, Class B, MBL

Graduated pipettes, type 2, Class B, MBL

1 ml, 0,01 ml, Yellow
Catalog # 612-3191
Supplier:   SciLabware
Graduated pipettes, type 2, Class B, MBL
Catalog # 612-3191
Supplier:  SciLabware
Graduated pipettes, type 2, Class B, MBL
Graduated pipettes, type 2, Class B, MBL

1 ml, 0,01 ml, Yellow

Price
18,80 €
Log in for availability
1 * 5 UN
Product Details & Documents

Soda lime glass.

  • Calibrated to deliver from any graduation line down to zero at the jet
  • Graduations in permanent amber stain

BS 700, ISO 835

Certifications: BS 700, ISO 835

Specifications
  • Colour code:
    Yellow
  • Capacity:
    1 ml
  • Division:
    0,01 ml