THERMOSTAT PROCESS INTEGRAL XT 750 H | -50 ... 300C | HEATING CAPACITY: 5,2 KW | COOLING CAPACITY: 6,7 KW 1 * 1 items
461-2941
: LAUDA
THERMOSTAT PROCESS INTEGRAL XT 750 H | -50 ... 300C | HEATING CAPACITY: 5,2 KW | COOLING CAPACITY: 6,7 KW 1 * 1 items
461-2941
: LAUDA
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection