Imaging System Gel Doc-It TS GelCam camera 310 (manual zoom lens): 12 bit monochrome - 16 File Bit Depth (A/D) digital camera with mount - 1600 x 1200 chip 1 * 1 items
Catalog # 733-2015
Supplier: UVP ULTRA VIOLET PRODUCTS
Frequently Bought Together