Bromophenol blue
Glass bottle for solids, 50 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 115-39-9 Molecular Formula: C19H10Br4O5S Formula Weight: 669.98 Odour: Odourless Melting Point: ca 270° dec. Beilstein Reference: 61698 Merck Reference: 14,1444 MDL No.: MFCD00005875
Synonyms: Bromphenol Blue Sultone Form, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein
- CAS Number:115-39-9
- Formula:C₁₉H₁₀Br₄O₅S
- MDL Number:MFCD00005875
- Molecular Weight:669.97 g/mol
- Merck Index:14,1444
- Melting Pt:∼ 270 °C (dec.)
- Density:1 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:204-086-2
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



