Ammonium acetate ≥97%
Plastic bottle, 250 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 97% CAS No.: 631-61-8 Molecular Formula: C2H7NO2 Formula Weight: 77.08 Flash Point: 136°(277°F) Odour: Ammonia-like Boiling Point: dec. Beilstein Reference: 4186741 Merck Reference: 14,495 MDL No.: MFCD00013066
Synonyms: Acetic acid ammonium salt
- CAS Number:631-61-8
- Formula:H₃CCOONH₄
- MDL Number:MFCD00013066
- Molecular Weight:77.08 g/mol
- Merck Index:14,495
- Boiling Pt:Dec.
- Melting Pt:∼114 °C (dec.)
- Density:1.17
- Flash Pt:136 °C (277 °F)
- Storage Temperature:Ambient
- EINECS:211-162-9
Documents
Specifications
Specification Test Results
- Form:Crystals or powder or crystalline powder
- Identification (FTIR):Conforms
- Ammonia as ammonium acetate:≥96.0% to ≤104.0%
- Water Content (Karl Fischer Titration):≤5.0%
- Appearance (Color):White
Specifications
- Pack type:Plastic bottle
- Pk:250 g



