Order Entry
Spain
Orders LinkContactUsLinkComponent
3-Octanone ≥98%

3-Octanone ≥98%

Glass bottle for solids, 25 g
Catalog # A12113.14
Flammable
Irritant
3-Octanone ≥98%
Catalog # A12113.14
CAS Number:  106-68-3
3-Octanone ≥98%
3-Octanone ≥98%

Glass bottle for solids, 25 g

CAS Number:  106-68-3
Price
39,10 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98% CAS No.: 106-68-3 Molecular Formula: C8H16O Formula Weight: 128.22 Flash Point: 51°(123°F) Refractive Index: 1.4150 Melting Point: -23° Boiling Point: 167-168° Beilstein Reference: 1700021 Merck Reference: 14,6084 UN No.: 1224 MDL No.: MFCD00009515

  • CAS Number:106-68-3
  • Formula:C₈H₁₆O
  • MDL Number:MFCD00009515
  • Molecular Weight:128.21 g/mol
  • Merck Index:14,6084
  • Boiling Pt:167…168 °C
  • Melting Pt:–23 °C
  • Density:0.822
  • Flash Pt:51 °C (123 °F)
  • Storage Temperature:Ambient
  • EINECS:203-423-0
  • UN:1224
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications