Order Entry
Spain
ContactUsLinkComponent
Pipette volumetric, capacity 25ml, class As, amber graduation, soda-lime glass, tolerance 0.03ml, total length +-10mm:520mm, conform ity certificate. 1 * 1 items
Catalog # HIRS134017037
Supplier:  HIRSCHMANN
undefined
Pipette volumetric, capacity 25ml, class As, amber graduation, soda-lime glass, tolerance 0.03ml, total length +-10mm:520mm, conform ity certificate. 1 * 1 items
Catalog # HIRS134017037
Supplier:  HIRSCHMANN
Frequently Bought Together