Order Entry
Spain
Orders LinkContactUsLinkComponent
Wall - mounted cabinets
Wall - mounted cabinets
Wall-mounted cabinet, with 6 shelves, 2, 920×300×800 mm
Catalog # 132-0401
Supplier:   Palbam
Wall - mounted cabinets
Catalog # 132-0401
Supplier:  Palbam
Wall - mounted cabinets
Wall - mounted cabinets
Wall-mounted cabinet, with 6 shelves, 2, 920×300×800 mm
Price
3 310,00 €
Log in for availability
1 * 1 UN
Product Details & Documents
Stainless steel 304 with electropolished finish
  • High-quality finish
Packaging: Typically available from stock for direct shipment from the supplier. Shipped flat-packed for simple assembly by the customer. Assembly instructions included.
Specifications
  • Description:
    Wall-mounted cabinet, with 6 shelves
  • W×D×H:
    920×300×800 mm
  • Doors:
    2