Order Entry
Spain
ContactUsLinkComponent
Flask volumetric, capacity 1l, class A, amber graduated, glass stoppered, tolerance 0.4ml, height 300mm, NS-ground 24/29, graduation color white, conformity certified. 1 * 1 items
Catalog # HIRS2640293_S
Supplier:  HIRSCHMANN
undefined
Flask volumetric, capacity 1l, class A, amber graduated, glass stoppered, tolerance 0.4ml, height 300mm, NS-ground 24/29, graduation color white, conformity certified. 1 * 1 items
Catalog # HIRS2640293_S
Supplier:  HIRSCHMANN
Frequently Bought Together