Order Entry
Spain
ContactUsLinkComponent
Flask volumetric, capacity 500ml, class A, amber graduated, glass stoppered, tolerance 0.25ml, height 260mm, NS-ground 19/26, gradua tion color white, conformity certified. 1 * 1 items
Catalog # HIRS2640290_S
Supplier:  HIRSCHMANN
undefined
Flask volumetric, capacity 500ml, class A, amber graduated, glass stoppered, tolerance 0.25ml, height 260mm, NS-ground 19/26, gradua tion color white, conformity certified. 1 * 1 items
Catalog # HIRS2640290_S
Supplier:  HIRSCHMANN
Frequently Bought Together