Order Entry
Spain
ContactUsLinkComponent
Flask volumetric, capacity 10ml, class A, PE Stoppered, tolerance 0.025ml, height 90mm, NS-ground 7/16, graduation color blue, con f ormity certified. 1 * 2 items
Catalog # HIRS2820160_S
Supplier:  HIRSCHMANN
undefined
Flask volumetric, capacity 10ml, class A, PE Stoppered, tolerance 0.025ml, height 90mm, NS-ground 7/16, graduation color blue, con f ormity certified. 1 * 2 items
Catalog # HIRS2820160_S
Supplier:  HIRSCHMANN
Frequently Bought Together