Order Entry
Spain
ContactUsLinkComponent
Pipette volumetric, capacity 100ml, class AS, graduation brown in mass, tolerance 0.08ml, total length +-10mm:585mm, conformity cert ified. 1 * 1 items
Catalog # HIRS1340180_S
Supplier:  HIRSCHMANN
undefined
Pipette volumetric, capacity 100ml, class AS, graduation brown in mass, tolerance 0.08ml, total length +-10mm:585mm, conformity cert ified. 1 * 1 items
Catalog # HIRS1340180_S
Supplier:  HIRSCHMANN
Frequently Bought Together