Order Entry
Spain
ContactUsLinkComponent
Pipette volumetric, capacity 5ml, class AS, amber graduated, AR-glass, tolerance 0.015ml, total length +-10mm:400mmconformity certif ied. 1 * 1 items
Catalog # HIRS1340155_S
Supplier:  HIRSCHMANN
undefined
Pipette volumetric, capacity 5ml, class AS, amber graduated, AR-glass, tolerance 0.015ml, total length +-10mm:400mmconformity certif ied. 1 * 1 items
Catalog # HIRS1340155_S
Supplier:  HIRSCHMANN
Frequently Bought Together