Order Entry
Spain
ContactUsLinkComponent
Pipette volumetric, capacity 20ml, class AS, blue graduation, tolerance 0.03ml, total length +-10mm:510mmconformity certified. 1 * 1 items
Catalog # HIRS1341165_S
Supplier:  HIRSCHMANN
Pipette volumetric, capacity 20ml, class AS, blue graduation, tolerance 0.03ml, total length +-10mm:510mmconformity certified. 1 * 1 items
Catalog # HIRS1341165_S
Supplier:  HIRSCHMANN
Frequently Bought Together