Order Entry
Spain
Orders LinkContactUsLinkComponent
Capillary columns, select silanes

Capillary columns, select silanes

Column, silane, temp. limits 0 to 270/300 °C, Capillary, Select Silanes, 1,80 µm, 0,32 mm, 30 m
Catalog # VARICP7434
Supplier:   VARIAN
Capillary columns, select silanes
Catalog # VARICP7434
Supplier:  VARIAN
Capillary columns, select silanes

Column, silane, temp. limits 0 to 270/300 °C, Capillary, Select Silanes, 1,80 µm, 0,32 mm, 30 m

Price
1 452,66 €
Log in for availability
1 * 1 UN
Product Details & Documents

The Select Silanes column is a stabilized trifluoropropyl-methyl polysiloxane phase optimized for silanes determination. The Select Silanes column has a very thick film, resulting in high capacity and retention for highly volatile silanes.

  • High capacity and retention provide optimized productivity for silane analysis
  • Low bleed analysis to ppm levels for best detection limits and most accurate results
  • Reduced surface activity provides better peak shape for more reliable data
Specifications
  • Column Format:
    Capillary
  • Film thickness:
    1,80 µm
  • Ø int.:
    0,32 mm
  • Description:
    Column, silane, temp. limits 0 to 270/300 °C
  • Length:
    30 m
  • Seq:
    01
  • Phase:
    Select Silanes
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.