Order Entry
Spain
Orders LinkContactUsLinkComponent
 

Cefadroxil, Reference Standards

Cefadroxil, USP reference standard, Vial, 200 mg
  1097104.
 :   USP
Cefadroxil, Reference Standards
  1097104.
 :  USP
 :  
Cefadroxil, Reference Standards

Cefadroxil, USP reference standard, Vial, 200 mg

 :  
 
406,00 €
   
1 * 200 mg
 

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

 : US Pharmacopea Reference Standard

 

 

 

  • Pack type:
    Vial
  • Pk:
    200 mg
  • Standards:
    USP reference standard
  • Description:
    Cefadroxil
  • Cat. No.:
    1097104.
  • Formula:
    C₁₆H₁₇N₃O₅S
  • Description:
    Cefadroxil
  • CAS No.:
    66592-87-8