TICKOPUR R 32 - 2 LITRES, SPECIAL CLEANER, WITHOUT COMPLEXING AGENTS, WITH CORROSION PROTECTION, CONCENTRATE, GENTLE TO MATERIAL, DO SAGE 0.25 - 5 %, MILDLY ALKALINE, PH 9.9 (1 %) 1 * 2 L
Catalog # BAND882
Supplier: BANDELIN-ELECTRONIC
TICKOPUR R 32 - 2 LITRES, SPECIAL CLEANER, WITHOUT COMPLEXING AGENTS, WITH CORROSION PROTECTION, CONCENTRATE, GENTLE TO MATERIAL, DO SAGE 0.25 - 5 %, MILDLY ALKALINE, PH 9.9 (1 %) 1 * 2 L
Catalog # BAND882
Supplier: BANDELIN-ELECTRONIC
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Detergents, liquid, for automated cleaning of glassware, neodisher® FLA
Supplier: Dr. Weigert
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Water, TECHNICAL, demineralised
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
VWR® Beakers, Disposable
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals