Order Entry
Spain
ContactUsLinkComponent
TICKOPUR R 32 - 1 LITRE, SPECIAL CLEANER, WITHOUT COMPLEXING AGENTS, WITH CORROSION PROTECTION, CONCENTRATE, GENTLE TO MATERIAL, DOS AGE 0.25 - 5 %, MILDLY ALKALINE, PH 9.9 (1 %) 1 * 1 L
Catalog # BAND828
TICKOPUR R 32 - 1 LITRE, SPECIAL CLEANER, WITHOUT COMPLEXING AGENTS, WITH CORROSION PROTECTION, CONCENTRATE, GENTLE TO MATERIAL, DOS AGE 0.25 - 5 %, MILDLY ALKALINE, PH 9.9 (1 %) 1 * 1 L
Catalog # BAND828
Frequently Bought Together