Order Entry
Spain
Orders LinkContactUsLinkComponent
Microorganisms, E-power™
Microorganisms, E-power™
Candida albicans ATCC® 10231™, 10⁷ CFU per pellet, 10
Catalog # 0443E7
Marketsource
Supplier:   MICROBIOLOGICS
Microorganisms, E-power™
Catalog # 0443E7
Supplier:  MICROBIOLOGICS
Marketsource
CAS Number:  
Microorganisms, E-power™
Microorganisms, E-power™
Candida albicans ATCC® 10231™, 10⁷ CFU per pellet, 10
CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 KIT
Product Details & Documents
Epower™ microorganisms can be used to perform many essential quality control functions including microbial detection and enumeration, equipment calibration, method validation, bioburden determination, and antibacterial effectiveness and lethality testing. Available in concentrations ranging from 10² to 10⁸ CFU per pellet, Epower™ lyophilized microorganism preparations are extremely versatile. Epower™ pellets can be used individually or several Epower™ pellets can be combined for a mixed microorganism population challenge. They can also be easily manipulated to deliver a wide variety of concentrations. For added convenience, a peel-off Certificate of Assay is included for quality control documentation. Epower™ is packaged in a resealable vial containing 10 pellets of a single quantitative microorganism strain.
The ATCC licensed derivative emblem, the ATCC licensed derivative word mark and the ATCC catalog marks are trademarks of ATCC. Microbiologics, Inc. is licensed to use these trademarks and to sell products derived from ATCC® cultures.
  • Quantitative
  • Versatile
  • Does not require frozen storage conditions
  • Traceable to reference culture
Attention: Other microorganisms are available in the same format. Please contact Avantor to get more information.
Documents
Specifications
Specifications
  • Pk:
    10
  • Description:
    Candida albicans ATCC® 10231™, 10⁷ CFU per pellet