Order Entry
Spain
Orders LinkContactUsLinkComponent
Inlet liners

Inlet liners

Split/splitless, Siltek, with wool, universal for most common analyses, 5,0 mm, 3,5 mm, 95 mm
Catalog # 568-2351
Inlet liners
Catalog # 568-2351
Supplier:  
Inlet liners
Inlet liners

Split/splitless, Siltek, with wool, universal for most common analyses, 5,0 mm, 3,5 mm, 95 mm

Discontinued
Product Details & Documents

These liners can be used with Agilent GCs.

Ø int. = Nominal diameter at syringe needle expulsion point

Specifications
  • Ext.Ø:
    5,0 mm
  • Ø int.:
    3,5 mm
  • Description:
    Split/splitless, Siltek, with wool, universal for most common analyses
  • Length:
    95 mm
  • Seq:
    070
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.