Order Entry
Spain
Orders LinkContactUsLinkComponent
Inlet liners

Inlet liners

Drilled uniliner for trace, active samples, high recovery and linearity, 6,3 mm, 4,0 mm, 78,5 mm
  REST21055
 :   Restek
Inlet liners
  REST21055
 :  Restek
Inlet liners
Inlet liners

Drilled uniliner for trace, active samples, high recovery and linearity, 6,3 mm, 4,0 mm, 78,5 mm

 
433,00 €
   
1 * 5 UN
 

These liners can be used with Agilent GCs.

Ø int. = Nominal diameter at syringe needle expulsion point

 
  • Ext.Ø:
    6,3 mm
  • Ø int.:
    4,0 mm
  • Description:
    Drilled uniliner for trace, active samples, high recovery and linearity
  • Length:
    78,5 mm
  • Seq:
    040
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.