Capillary column caps
Capillary column caps, glass
Catalog # REST21044
Supplier: Restek
Jump to:
Product Details & Documents
These non-reusable column caps increase column lifetime, prevent moisture and air from entering the column during storage.
- Attach to the column in seconds to form an airtight seal
- Available in two styles, glass or silicone material
- Glass caps are colour coded for identifying detector and injector ends
Specifications
- Description:Capillary column caps, glass
- Seq:01