Order Entry
Spain
Orders LinkContactUsLinkComponent
Replacement inlet seals
Replacement inlet seals
Inlet seal, used with single column, gold plated, 0,8 mm Ø int.
Catalog # REST21318
Supplier:   Restek
Replacement inlet seals
Catalog # REST21318
Supplier:  Restek
Replacement inlet seals
Replacement inlet seals
Inlet seal, used with single column, gold plated, 0,8 mm Ø int.
Price
337,00 €
Log in for availability
1 * 10 UN
Product Details & Documents
Inlet seals are used at the base of the Agilent 5890/6890/7890 GC injection port, which contacts the sample and should be changed frequently to prevent adsorption of active compounds. In addition, septum fragments and sample residue accumulate on the disk surface, requiring disk replacement.
Specifications
  • Description:
    Inlet seal, used with single column, gold plated, 0,8 mm Ø int.
  • Seq:
    02
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Replacement inlet seal washers
    Replacement inlet seal washers
    REST21710
    Log in for availability
    1 * 15 UN - 17,00 €