Inlet liners
Splitless, double gooseneck, for trace, active samples up to 4 µl, for Varian 1075/1077, 6,3, 4,0 mm, 74 mm
Catalog # REST20849
Jump to:
Discontinued
Product Details & Documents
These liners can be used with Agilent GCs.
Ø int. = Nominal diameter at syringe needle expulsion point
Specifications
- Ext.Ø:6,3
- Ø int.:4,0 mm
- Description:Splitless, double gooseneck, for trace, active samples up to 4 µl, for Varian 1075/1077
- Length:74 mm
- Seq:061
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



