Order Entry
Spain
Orders LinkContactUsLinkComponent
 

Calibration weights, class F1

50 g, 0,30 ± mg, Cylindrical test weight
Catalog # SARBYCW4538-00
Supplier:   Sartorius Balances
Calibration weights, class F1
Catalog # SARBYCW4538-00
Supplier:  Sartorius Balances
Calibration weights, class F1
Calibration weights, class F1

50 g, 0,30 ± mg, Cylindrical test weight

Price
104,00 €
Log in for availability
1 * 1 UN
Product Details & Documents

Cylindrical weights which are made up of special stainless steel with a density of 7,950 kg/m³.

Certifications: Comply in all respects with the requirements on shape, material and markings specified by the OIML R111-1 International Recommendation.

Delivery information: Cylindrical weights are supplied in high-quality plastic cases that are suitable for use in cleanrooms.

Specifications
  • Weight:
    50 g
  • Seq:
    06
  • Tolerance:
    0,30 ± mg
  • Type:
    Cylindrical test weight
Recomendaciones

Las recomendaciones se personalizarán según sus preferencias de compra solo si ha dado su consentimiento activando la opción "Mejorar mi experiencia de compra" en la "página de información personal".

De lo contrario, recibirá recomendaciones genéricas.