Order Entry
Spain
Orders LinkContactUsLinkComponent
 

Inlet liners

Splitless, double gooseneck, for trace, active samples up to 4 µl, for Varian 1075/1077, 6,3 mm, 4,0 mm, 74
Catalog # REST20848
Inlet liners
Catalog # REST20848
Supplier:  
Inlet liners
Inlet liners

Splitless, double gooseneck, for trace, active samples up to 4 µl, for Varian 1075/1077, 6,3 mm, 4,0 mm, 74

Discontinued
Product Details & Documents

These liners can be used with Agilent GCs.

Ø int. = Nominal diameter at syringe needle expulsion point

Specifications
  • Ext.Ø:
    6,3 mm
  • Ø int.:
    4,0 mm
  • Description:
    Splitless, double gooseneck, for trace, active samples up to 4 µl, for Varian 1075/1077
  • Length:
    74
  • Seq:
    060