Order Entry
Spain
ContactUsLinkComponent
In vivo-jetpei-fluor is a fluorescently labeled dna and sirna delivery reagent for in vivo experiments in animal models ( tetramethy lrhodamine-conjugated, red label, excitation at 555 nm emission at 580 nm). 1 * 100 µl
Catalog # 206-10G
undefined
In vivo-jetpei-fluor is a fluorescently labeled dna and sirna delivery reagent for in vivo experiments in animal models ( tetramethy lrhodamine-conjugated, red label, excitation at 555 nm emission at 580 nm). 1 * 100 µl
Catalog # 206-10G