Syringes
Syringe, glass, metal centre nozzle, Luer slip, grad. 5,0 ml, 100 ml
Catalog # 613-4511
Supplier: MURRAY S
Jump to:
Product Details & Documents
Glass syringes with metal centre nozzle, without needles.
- Centre nozzle with Luer slip fitting
Specifications
- Volume:100 ml
- Description:Syringe, glass, metal centre nozzle, Luer slip, grad. 5,0 ml
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



