Order Entry
Spain
Orders LinkContactUsLinkComponent
 

Cesium chloride

Glass bottle for solids, 100 g
Catalog # AMBI9162
Marketsource
Supplier:   Ambion
Cesium chloride
Catalog # AMBI9162
Supplier:  Ambion
Marketsource
CAS Number:  7647-17-8
Cesium chloride

Glass bottle for solids, 100 g

CAS Number:  7647-17-8
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Cesium Chloride
  • CAS Number:7647-17-8
  • Formula:CsCl
  • MDL Number:MFCD00010955
  • Molecular Weight:168.36 g/mol
  • Merck Index:13,02021
  • Boiling Pt:1303 °C (1013 hPa)
  • Melting Pt:646 °C
  • Density:3.7808 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-600-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.