Reference standards VOA
m-xylene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule, 1 mL
Catalog # REST30424
Supplier: Restek
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:1 mL
- Description:m-xylene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- Cat. No.:REST30424
- Formula:C₈H₁₀
- Description:m-xylene
- CAS No.:108-38-3



