Order Entry
Spain
Orders LinkContactUsLinkComponent
Reference standards VOA

Reference standards VOA

1,2,3-trichlorobenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule, 1 mL
Catalog # REST30416
Supplier:   Restek
Reference standards VOA
Catalog # REST30416
Supplier:  Restek
CAS Number:  
Reference standards VOA

1,2,3-trichlorobenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule, 1 mL

CAS Number:  
Price
53,00 €
Log in for availability
1 * 1 mL
Product Details & Documents
Specifications

Specifications

  • Pk:
    1 mL
  • Description:
    1,2,3-trichlorobenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
  • Cat. No.:
    REST30416
  • Formula:
    C₆H₃Cl₃
  • Description:
    1,2,3-trichlorobenzene
  • CAS No.:
    87-61-6

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.