Reference standards VOA
Tetrahydrofuran, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule, 1
Catalog # REST30414
Supplier: Restek
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:1
- Description:Tetrahydrofuran, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- Cat. No.:REST30414
- Formula:C₄H₈O
- Description:Tetrahydrofuran
- CAS No.:109-99-9



