Reference standards VOA
Pyridine, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule, 1
Catalog # REST30409
Supplier: Restek
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:1
- Description:Pyridine, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- Cat. No.:REST30409
- Formula:C₅H₅N
- Description:Pyridine
- CAS No.:110-86-1



