Reference standards VOA
Isopropylbenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule, 1 mL
Catalog # REST30294
Supplier: Restek
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:1 mL
- Description:Isopropylbenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- Description:Isopropylbenzene
- CAS No.:98-82-8
- Cat. No.:REST30294
- Formula:C₉H₁₂



