Reference standards VOA
Isobutyl alcohol, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule, 1
Catalog # REST30293
Supplier: Restek
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:1
- Description:Isobutyl alcohol, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- Cat. No.:REST30293
- Formula:C₄H₁₀O
- Description:Isobutyl alcohol
- CAS No.:78-83-1



